Anti-MRPL52 Rabbit Polyclonal Antibody
Catalog # 77701-780
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MRPL52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q86TS9
- Antigen synonyms:MRP-L52|39S ribosomal protein L52|mitochondrial|Mrpl52|RM52_HUMAN|L52mt
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human MRP-L52.
- Purification:Antigen affinity purified
- Cat. no.:77701-780
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:5000
Type:
Antigen: MRPL52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human