Anti-CYP2R1 Rabbit Polyclonal Antibody
77701-761
New Product
- Antigen symbol:CYP2R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q6VVX0
- Antigen synonyms:CYP2R1|Cytochrome P450 family 2 R1|Vitamin D 25 hydroxylase|Cytochrome P450 2R1|Cytochrome P450 family 2 subfamily R polypeptide 1|CP2R1_HUMAN|Vitamin D 25-hydroxylase|EC 1.14.13.15|MGC4663
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CYP2R1.
- Purification:Antigen affinity purified
- Cat. no.:77701-761
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: CYP2R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Monkey