Anti-VPS26B Rabbit Polyclonal Antibody
Catalogue # 77701-713
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:VPS26B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q4G0F5
- Antigen synonyms:Vacuolar protein sorting-associated protein 26B|Pep8b|Vacuolar protein sorting 26 homolog B (S. pombe)|VP26B_HUMAN|VPS26B|Vacuolar protein sorting 26 homolog B (yeast)|Vesicle protein sorting 26B
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human VPS26B.
- Purification:Antigen affinity purified
- Cat. no.:77701-713
- Packaging:Vial
Spécifications
À propos de cet article
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: VPS26B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse