Anti-ARPP21 Rabbit Polyclonal Antibody
Catalog # 77701-635
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ARPP21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UBL0
- Antigen synonyms:cAMP-regulated phosphoprotein|OTTHUMP00000161358|Ppp1r1c|ARPP21|OTTHUMP00000208638|OTTHUMP00000208640|Tarpp|R3H domain containing 3|21|21kDa|Cyclic AMP-regulated phosphoprotein|OTTHUMP00000208657|RCS|ARP21_HUMAN|OTTHUMP00000208653|FLJ32997|MGC109368|RGD1307208|OTTHUMP00000208651|Ppp1r1cl|RGD1307215|21-KD|cAMP-regulated phosphoprotein 21|OTTHUMP00000208639|R3HDM3|OTTHUMP00000208652|Regulator of calmodulin signaling|Thymocyte cAMP-regulated phosphoprotein|OTTHUMP00000208641|cAMP regulated phosphoprotein|0710001E13Rik|AI853636|OTTHUMP00000208648|OTTHUMP00000208646|ARPP-21|D9Bwg1012e|OTTHUMP00000208635|OTTHUMP00000208636|OTTHUMP00000208637
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ARPP-21.
- Purification:Antigen affinity purified
- Cat. no.:77701-635
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ARPP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse