Anti-AKAP3 Rabbit Polyclonal Antibody
Catalog # 77701-589
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:AKAP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:O75969
- Antigen synonyms:AKAP-3|FSP95|PRKA3|Sperm oocyte binding protein 1|Fibrousheathin-1|Protein kinase A-anchoring protein 3|Akap3|AKAP 110|Fibrousheathin I|Sperm oocyte-binding protein|protein kinase A binding protein AKAP 110|A kinase (PRKA) anchor protein 3|Fibrousheathin 1|SOB 1|A-kinase anchor protein 110 kDa|A-kinase anchor protein 3|Cancer/testis antigen 82|AKAP110|AKAP3_HUMAN|SOB1|CT82|Fibrous sheath protein of 95 kDa
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human AKAP 110.
- Purification:Antigen affinity purified
- Cat. no.:77701-589
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AKAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse