Anti-NCOA2 Rabbit Polyclonal Antibody
77701-569
New Product
- Antigen symbol:NCOA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q15596
- Antigen synonyms:TIF 2|Oncogene FGR|NCoA-2|hTIF2|TIF2|SRC 2|Nuclear receptor coactivator 2|glucocorticoid receptor interacting protein 1|NCoA 2|Transcriptional intermediary factor 2|src2|NCOA2_HUMAN|bHLHe75|p160|MED1|GRIP 1|GRIP1|Ncoa2|Steroid receptor coactivator 2|Class E basic helix-loop-helix protein 75|MGC138808
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human GRIP-1 around the non-phosphorylation site of S736.
- Purification:Antigen affinity purified
- Cat. no.:77701-569
- Packaging:Vial
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: NCOA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human