Anti-MERTK/TYRO3 (phospho-Y749/681) Rabbit Polyclonal Antibody
Catalog # 77701-560
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MERTK/TYRO3 (phospho-Y749/681)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q12866/Q06418
- Antigen synonyms:TYRO3|RSE|MERTK|Tyrosine-protein kinase RSE|DTK|SKY|Receptor tyrosine kinase MerTK|Tyrosine-protein kinase receptor TYRO3|Tyrosine-protein kin|Proto-oncogene c-Mer|Tyrosine-protein kinase Mer|MER|Tyrosine-protein kinase DTK|BYK
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human MerTK/Tyro3 around the phosphorylation site of Y749/681.
- Purification:Antigen affinity purified
- Cat. no.:77701-560
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: MERTK/TYRO3 (phospho-Y749/681)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat