Anti-CRYBB1 Rabbit Polyclonal Antibody
Catalog # 77687-459
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CRYBB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P53674
- Antigen synonyms:Beta-B1 crystallin|Crystallin beta B1|CRYBB 1|CRBB1_HUMAN|CATCN3|Beta crystallin B1|Crybb1|OTTHUMP00000028719|CTRCT17|Eye lens structural protein|Beta-crystallin B1
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-crystallin B1 protein (1-252AA)
- Purification:Antigen affinity purified
- Cat. no.:77687-459
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: CRYBB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse