Anti-CRTAC1 Rabbit Polyclonal Antibody
Catalogue # 77687-454
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:CRTAC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NQ79
- Antigen synonyms:68 kDa chondrocyte expressed protein|Chondrocyte expressed protein 68 kDa CEP 68|Protein CRTAC1-B|68 kDa chondrocyte-expressed protein|Cartilage acidic protein 1|CEP-68|Acidic secreted protein in cartilage|ASPIC1|CRTAC1|CEP 68 |CRAC1_HUMAN|ASPIC|CEP68 |Cartilage acidic protein 1 precursor|W307
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cartilage acidic protein 1 protein (309-661AA)
- Purification:Antigen affinity purified
- Cat. no.:77687-454
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB:1:500-1:2000
Type:
Antigen: CRTAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human