Anti-mpt64 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: mpt64
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium tuberculosis
Specifications
- Catalog No:
- 77710-237
- 77710-236
- Antigen synonyms:
- Rv1980c|Immunogenic protein MPT64|MTCY39.39|mpt64|Antigen MPT64
- Rv1980c|Immunogenic protein MPT64|MTCY39.39|mpt64|Antigen MPT64
- Gene ID:
- P9WIN9
- P9WIN9
- Immunogen:
- Recombinant Mycobacterium tuberculosis Immunogenic protein MPT64 protein (24-228AA)
- Recombinant Mycobacterium tuberculosis Immunogenic protein MPT64 protein (24-228AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together