Anti-CLDND1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: CLDND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77709-891
- 77709-890
- Antigen synonyms:
- UNQ2511/PRO6000Claudin domain-containing protein 1|PSEC0054|Membrane protein GENX-3745|CLDND1|HSPC174|C3orf4
- UNQ2511/PRO6000Claudin domain-containing protein 1|PSEC0054|Membrane protein GENX-3745|CLDND1|HSPC174|C3orf4
- Gene ID:
- Q9NY35
- Q9NY35
- Immunogen:
- Fusion protein of Human CLDND1
- Fusion protein of Human CLDND1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together