Anti-INHBB Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: INHBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77689-183
- 77709-531
- Antigen synonyms:
- Inhibin beta 2|INHBB|Activin beta B chain|Inhibin beta B subunit|Inhibin beta B chain|Activin beta-B chain|INHBB_HUMAN
- Inhibin beta 2|INHBB|Activin beta B chain|Inhibin beta B subunit|Inhibin beta B chain|Activin beta-B chain|INHBB_HUMAN
- Gene ID:
- P09529
- P09529
- Immunogen:
- Recombinant Human Inhibin beta B chain protein (295-405AA)
- Recombinant Human Inhibin beta B chain protein (295-405AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together