Anti-ANAPC4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:50-1:200
Type:
Antigen: ANAPC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77709-299
- 77709-298
- Antigen synonyms:
- APC Complex Protein CUT20|Anaphase Promoting Complex Subunit 4|APC4|CUT20|APC4_HUMAN|ANAPC 4|Cyclosome subunit 4|APC 4|LID1|anapc4|Anaphase-promoting complex subunit 4
- APC Complex Protein CUT20|Anaphase Promoting Complex Subunit 4|APC4|CUT20|APC4_HUMAN|ANAPC 4|Cyclosome subunit 4|APC 4|LID1|anapc4|Anaphase-promoting complex subunit 4
- Gene ID:
- Q9UJX5
- Q9UJX5
- Immunogen:
- Synthetic peptide of Human ANAPC4
- Synthetic peptide of Human ANAPC4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together