Anti-PIK3CG Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIK3CG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77709-166
- 77709-165
- Antigen synonyms:
- PtdIns-3-kinase subunit gamma|p110 gamma|Phosphoinositide 3 kinase catalytic gamma polypeptide|Serine/threonine protein kinase PIK3CG|PI3-kinase subunit gamma|PI3CG|Phosphatidylinositol 4 5 bisphosphate 3 kinase 110 kDa catalytic subunit gamma|PI3K|Phosphatidylinositol 3 kinase, catalytic, gamma polypeptide|Phosphatidylinositol 3 kinase catalytic 110 kD gamma|PtdIns-3-kinase subunit p110-gamma|PI3Kgamma|Phosphatidylinositol 3 kinase gamma, p110 gamma|Phosphoinositide 3 kinase gamma catalytic subunit|5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma|PI3K-gamma|p110gamma|5-bisphosphate 3-kinase catalytic subunit gamma isoform|p120 PI3K|Phosphatidylinositol 4 5 bisphosphate 3 kinase catalytic subunit gamma isoform|Phosphatidylinositol 4 5 bisphosphate 3 kinase catalytic subunit gamma|Pik3cg|PI 3 Kinase catalytic subunit gamma|1 phosphatidylinositol 3 kinase|PIK3|p120-PI3K|Phosphatidylinositol-4|PK3CG_HUMAN|PI3 kinase p110 subunit gamma
- PtdIns-3-kinase subunit gamma|p110 gamma|Phosphoinositide 3 kinase catalytic gamma polypeptide|Serine/threonine protein kinase PIK3CG|PI3-kinase subunit gamma|PI3CG|Phosphatidylinositol 4 5 bisphosphate 3 kinase 110 kDa catalytic subunit gamma|PI3K|Phosphatidylinositol 3 kinase, catalytic, gamma polypeptide|Phosphatidylinositol 3 kinase catalytic 110 kD gamma|PtdIns-3-kinase subunit p110-gamma|PI3Kgamma|Phosphatidylinositol 3 kinase gamma, p110 gamma|Phosphoinositide 3 kinase gamma catalytic subunit|5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma|PI3K-gamma|p110gamma|5-bisphosphate 3-kinase catalytic subunit gamma isoform|p120 PI3K|Phosphatidylinositol 4 5 bisphosphate 3 kinase catalytic subunit gamma isoform|Phosphatidylinositol 4 5 bisphosphate 3 kinase catalytic subunit gamma|Pik3cg|PI 3 Kinase catalytic subunit gamma|1 phosphatidylinositol 3 kinase|PIK3|p120-PI3K|Phosphatidylinositol-4|PK3CG_HUMAN|PI3 kinase p110 subunit gamma
- Gene ID:
- P48736
- P48736
- Immunogen:
- Recombinant Human Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit gamma isoform protein (438-588AA)
- Recombinant Human Phosphatidylinositol 4,5-bisphosphate 3-kinase catalytic subunit gamma isoform protein (438-588AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together