Anti-FZD3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:25-1:100
Type:
Antigen: FZD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77708-924
- 77708-925
- Antigen synonyms:
- FZD3_HUMAN|FZD 3|Frizzled-3|Fzd3|Frizzled 3 seven transmembrane spanning receptor|Fz 3|Frizzled family receptor 3|Fz-3|Frizzled 3|Frizzled homolog 3|hFz 3|Frizzled3|Fz3|hFz3|Frizzled homolog 3 (Drosophila)
- FZD3_HUMAN|FZD 3|Frizzled-3|Fzd3|Frizzled 3 seven transmembrane spanning receptor|Fz 3|Frizzled family receptor 3|Fz-3|Frizzled 3|Frizzled homolog 3|hFz 3|Frizzled3|Fz3|hFz3|Frizzled homolog 3 (Drosophila)
- Gene ID:
- Q9NPG1
- Q9NPG1
- Immunogen:
- Synthetic peptide of Human FZD3
- Synthetic peptide of Human FZD3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together