Order Entry
Canada
ContactUsLinkComponent
Anti-RASD2 Rabbit Polyclonal Antibody
Anti-RASD2 Rabbit Polyclonal Antibody
Catalog # 77702-415
Supplier:  AFG Bioscience
New Product
undefined
Anti-RASD2 Rabbit Polyclonal Antibody
Catalog # 77702-415
Supplier:  AFG Bioscience
Supplier Number:  A21571-100UG
New Product

Specifications

  • Form:
    Liquid
  • Gene ID:
    Q96D21
  • Antigen synonyms:
    Tumor endothelial marker 2|TEM 2|RASD2|OTTHUMP00000197925|RASD 2|GTP-binding protein Rhes|Ras homolog enriched in striatum|RHES_HUMAN|RASD family member 2|GTP binding protein Rhes|Rhes|MGC:4834|TEM2
  • Storage buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
  • Storage temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synthesized peptide derived from the C-terminal region of Human Dexras2.
  • Purification:
    Antigen affinity purified
  • Cat. no.:
    77702-415
  • Packaging:
    Vial

Specifications

About this item

WB:1:500-1:2000, ELISA:1:40000

Type:
Antigen: RASD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat