Anti-PDZD2 Rabbit Polyclonal Antibody
Catalog # 77702-355
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:O15018
- Antigen synonyms:PDZK3|PDZD2|PDZ domain-containing protein 2|PDZ domain-containing protein 3) [Cleaved into: Processed PDZ domain-containing protein 2]|AIPC|Activated in prostate cancer protein|KIAA0300
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human PDZK3.
- Purification:Antigen affinity purified
- Cat. no.:77702-355
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PDZD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat