Anti-ZHX2 Rabbit Polyclonal Antibody
Catalog # 77702-326
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:Q9Y6X8
- Antigen synonyms:ZHX2|ZHX2_HUMAN|Alpha-fetoprotein regulator 1|Zinc finger and homeodomain protein 2|AFR1|Alpha fetoprotein regulator 1|RAF|Zinc fingers and homeoboxes 2|KIAA0854|Transcription factor ZHX2|Zinc fingers and homeoboxes protein 2|AFP regulator 1|Regulator of AFP
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human ZHX2.
- Purification:Antigen affinity purified
- Cat. no.:77702-326
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: ZHX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse