Anti-DES (phospho-S60) Rabbit Polyclonal Antibody
Catalog # 77702-061
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:P17661
- Antigen synonyms:CSM1|FLJ41013|CMD1I|DESM_HUMAN|DES|Desmin|OTTHUMP00000064865|Intermediate filament protein|FLJ12025|FLJ39719|FLJ41793|CSM2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Desmin around the phosphorylation site of S60.
- Purification:Antigen affinity purified
- Cat. no.:77702-061
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: DES (phospho-S60)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat