Anti-CYP11A1 Rabbit Polyclonal Antibody
Catalog # 77702-018
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:P05108
- Antigen synonyms:Cholesterol monooxygenase (side chain cleaving)|CYPXIA1|Cytochrome P450C11A1|Cytochrome P450 11A1|P450SCC|Cholesterol side-chain cleavage enzyme|Cytochrome P450(scc)|Cholesterol side chain cleavage enzyme mitochondrial|mitochondrial|Cytochrome P450 subfamily XIA|Cholesterol side chain cleavage enzyme|CP11A_HUMAN|Steroid 20 22 lyase|CYP11A|Cytochrome P450 11A1 mitochondrial|Cholesterol 20 22 desmolase|CYP11A1|Cholesterol desmolase|Cytochrome P450 family 11 subfamily A polypeptide 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human CYP11A1.
- Purification:Antigen affinity purified
- Cat. no.:77702-018
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:5000
Type:
Antigen: CYP11A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human