Anti-ECHS1 Rabbit Polyclonal Antibody
Catalog # 77687-556
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Gene ID:P30084
- Antigen synonyms:EC 4.2.1.17|Enoyl-CoA hydratase 1|mitochondrial|ECHS1Enoyl-CoA hydratase|Short-chain enoyl-CoA hydratase|SCEH
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Enoyl-CoA hydratase, mitochondrial protein (28-290AA)
- Purification:Antigen affinity purified
- Cat. no.:77687-556
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: ECHS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat