Order Entry
Canada
ContactUsLinkComponent
Anti-CDK5RAP2 Rabbit Polyclonal Antibody (Biotin)
Anti-CDK5RAP2 Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CDK5RAP2 Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77724-048
  • 77724-047
  • Antigen symbol:
  • CDK5RAP2
  • CDK5RAP2
  • Antigen synonyms:
  • CDK5RAP2|CDK5 regulatory subunit-associated protein 2|CDK5 regulatory subunit associated protein 2|C48|CEP215|MCPH3|Centrosome-associated protein 215|CDK5 activator-binding protein C48|centrosomal protein 215 kDa|CK5P2_HUMAN
  • CDK5RAP2|CDK5 regulatory subunit-associated protein 2|CDK5 regulatory subunit associated protein 2|C48|CEP215|MCPH3|Centrosome-associated protein 215|CDK5 activator-binding protein C48|centrosomal protein 215 kDa|CK5P2_HUMAN
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q96SN8
  • Q96SN8
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human CDK5 regulatory subunit-associated protein 2 protein (996-1111AA)
  • Recombinant Human CDK5 regulatory subunit-associated protein 2 protein (996-1111AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

Specifications

Frequently Bought Together