Anti-CAMK1G Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: CAMK1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77690-369
- 77723-927
- Antigen symbol:
- CAMK1G
- CAMK1G
- Antigen synonyms:
- VWS1|CLICK III|CaMK like CREB kinase III|CaM-KI gamma|Calcium/calmodulin-dependent protein kinase I-gamma|Camk1g|CaMKIG|CaM kinase I gamma|KCC1G_HUMAN|CaM kinase IG|CaMK-like CREB kinase III|Calcium/calmodulin dependent protein kinase type 1G|CLICK3|dJ272L16.1|CaMKI gamma|Calcium/calmodulin-dependent protein kinase Ig|Calcium/calmodulin-dependent protein kinase type 1G
- VWS1|CLICK III|CaMK like CREB kinase III|CaM-KI gamma|Calcium/calmodulin-dependent protein kinase I-gamma|Camk1g|CaMKIG|CaM kinase I gamma|KCC1G_HUMAN|CaM kinase IG|CaMK-like CREB kinase III|Calcium/calmodulin dependent protein kinase type 1G|CLICK3|dJ272L16.1|CaMKI gamma|Calcium/calmodulin-dependent protein kinase Ig|Calcium/calmodulin-dependent protein kinase type 1G
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96NX5
- Q96NX5
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Calcium/calmodulin-dependent protein kinase type 1G protein (217-476AA)
- Recombinant Human Calcium/calmodulin-dependent protein kinase type 1G protein (217-476AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together