Anti-CNNM3 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: CNNM3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77723-199
- 77723-198
- Antigen symbol:
- CNNM3
- CNNM3
- Antigen synonyms:
- ACDP3Metal transporter CNNM3|Ancient conserved domain-containing protein 3|Cyclin-M3|CNNM3
- ACDP3Metal transporter CNNM3|Ancient conserved domain-containing protein 3|Cyclin-M3|CNNM3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NE01
- Q8NE01
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Metal transporter CNNM3 protein (358-645AA)
- Recombinant Human Metal transporter CNNM3 protein (358-645AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together