Anti-ZDHHC15 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZDHHC15
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-811
- 77722-812
- Antigen symbol:
- ZDHHC15
- ZDHHC15
- Antigen synonyms:
- UNQ1969/PRO4501|EC 2.3.1.225|Zinc finger DHHC domain-containing protein 15|DHHC-15|ZDHHC15|Palmitoyltransferase ZDHHC15
- UNQ1969/PRO4501|EC 2.3.1.225|Zinc finger DHHC domain-containing protein 15|DHHC-15|ZDHHC15|Palmitoyltransferase ZDHHC15
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96MV8
- Q96MV8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Palmitoyltransferase ZDHHC15 protein (239-337AA)
- Recombinant Human Palmitoyltransferase ZDHHC15 protein (239-337AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together