Anti-SNX27 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SNX27
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-748
- 77722-747
- Antigen symbol:
- SNX27
- SNX27
- Antigen synonyms:
- MGC126871|Methamphetamine responsive transcript 1|MGC20471|SNX27|OTTHUMP00000042487|MRT1|Sorting nexin-27|SNX27_HUMAN|MGC126873|RP11-98D18.12|KIAA0488|Sorting nexin family member 27|MY014
- MGC126871|Methamphetamine responsive transcript 1|MGC20471|SNX27|OTTHUMP00000042487|MRT1|Sorting nexin-27|SNX27_HUMAN|MGC126873|RP11-98D18.12|KIAA0488|Sorting nexin family member 27|MY014
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96L92
- Q96L92
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sorting nexin-27 protein (458-497AA)
- Recombinant Human Sorting nexin-27 protein (458-497AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together