Anti-ELMOD3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:50-1:200
Type:
Antigen: ELMOD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-666
- 77690-302
- Antigen symbol:
- ELMOD3
- ELMOD3
- Antigen synonyms:
- RBED1|ELMOD3|RNA-binding protein 29|PP4068ELMO domain-containing protein 3|RNA-binding motif and ELMO domain-containing protein 1|RNA-binding motif protein 29|RBM29
- RBED1|ELMOD3|RNA-binding protein 29|PP4068ELMO domain-containing protein 3|RNA-binding motif and ELMO domain-containing protein 1|RNA-binding motif protein 29|RBM29
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96FG2
- Q96FG2
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human ELMO domain-containing protein 3 protein (1-102AA)
- Recombinant Human ELMO domain-containing protein 3 protein (1-102AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together