Anti-EIF2S2 (phospho–Ser2) Rabbit Polyclonal Antibody
Catalog # 77716-162
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:EIF2S2 (phospho-Ser2)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μl
- Form:liquid
- Gene ID:P20042
- Antigen synonyms:38kDa|PPP1R67|eIF 2 beta|EIF2|eukaryotic translation initiation factor 2|eukaryotic initiation factor 2-beta|EIF2S2|IF2B_HUMAN|subunit 2 beta|EIF2beta|EIF2B|Eukaryotic translation initiation factor 2 subunit beta|regulatory subunit 67|protein phosphatase 1|Eukaryotic initiation factor 2 beta|Eukaryotic translation initiation factor 2 subunit 2|Eukaryotic translation initiation factor 2 subunit 2 beta|eIF-2-beta|MGC8508|Eukaryotic translation initiation factor 2 beta|Eukaryotic translation initiation factor 2 subunit 2 beta 38kDa|DKFZp686L18198
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Peptide sequence around phosphorylation site of serine 2 (M-S(p)-G-D-E) derived from Human eIF2?.
- Purification:Antigen affinity purified
- Cat. no.:77716-162
- Packaging:Vial
Specifications
About this item
WB:1:500-1:1000
Type:
Antigen: EIF2S2 (phospho–Ser2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse