Anti-LYSMD3 Rabbit Polyclonal Antibody
Catalog # 77689-951
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:LYSMD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:Q7Z3D4
- Antigen synonyms:BC003322|lysM and putative peptidoglycan binding domain containing protein 3|LysM and putative peptidoglycan-binding domain-containing protein 3|RGD1308805|lysmd3|1110030H10Rik|LYSM3_HUMAN
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human LysM and putative peptidoglycan-binding domain-containing protein 3 protein (1-127AA)
- Purification:>95%, Protein G purified
- Cat. no.:77689-951
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: LYSMD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse