Anti-ZRSR2 Rabbit Polyclonal Antibody
77689-565
New Product
- Antigen symbol:ZRSR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:Q15696
- Antigen synonyms:CCCH domain-|CCCH type zinc finger|U2AF1RS2|U2AF1 RS2|U2AF1L2|MGC142014|U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit related protein 2|U2(RNU2) small nuclear RNA auxiliary factor 1-like 2|Renal carcinoma antigen NY-REN-20|URP|MGC142040|Zinc finger-|U2(RNU2) small nuclear RNA auxiliary factor 1 like 2|U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2|RNA binding motif and serine/arginine rich protein 2|Zrsr2|and RNA-binding motif-containing serine/arginine-rich protein 2|RNA-binding motif and serine/arginine rich protein 2|U2AF35-related protein|Renal carcinoma antigen NY REN 20|U2AFM_HUMAN
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2 protein (191-372AA)
- Purification:>95%, Protein G purified
- Cat. no.:77689-565
- Packaging:Vial
Type:
Antigen: ZRSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse