Anti-PPP2R3A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPP2R3A
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77713-553
- 77713-554
- Antigen symbol:
- PPP2R3A
- PPP2R3A
- Antigen synonyms:
- PR59|Protein phosphatase 2 (formerly 2A) regulatory subunit B'' alpha|Serine/threonine protein phosphatase 2A 72/130 kDa regulatory subunit B|PP2A subunit B isoforms B72/B130|PR72|PP2A subunit B B72/B130 isoforms|PP2A subunit B B''-PR72/PR130|Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha|PP2A subunit B R3 isoform|Ppp2r3a|Serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B|PR130|DNA for thyroid hormone receptor binding site (258bp)|P2R3A_HUMAN|PP2A subunit B isoforms B''-PR72/PR130|PP2A subunit B isoform PR72/PR130|PP2A subunit B PR72/PR130 isoforms|PPP2R3|PP2A subunit B isoform R3 isoform|AI118493
- PR59|Protein phosphatase 2 (formerly 2A) regulatory subunit B'' alpha|Serine/threonine protein phosphatase 2A 72/130 kDa regulatory subunit B|PP2A subunit B isoforms B72/B130|PR72|PP2A subunit B B72/B130 isoforms|PP2A subunit B B''-PR72/PR130|Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha|PP2A subunit B R3 isoform|Ppp2r3a|Serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B|PR130|DNA for thyroid hormone receptor binding site (258bp)|P2R3A_HUMAN|PP2A subunit B isoforms B''-PR72/PR130|PP2A subunit B isoform PR72/PR130|PP2A subunit B PR72/PR130 isoforms|PPP2R3|PP2A subunit B isoform R3 isoform|AI118493
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q06190
- Q06190
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Serine/threonine-protein phosphatase 2A regulatory subunit B\'\' subunit alpha protein (256-508AA)
- Recombinant Human Serine/threonine-protein phosphatase 2A regulatory subunit B\'\' subunit alpha protein (256-508AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together