Order Entry
Canada
ContactUsLinkComponent
Anti-TAF11 Rabbit Polyclonal Antibody
Anti-TAF11 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-TAF11 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77713-489
  • 77713-488
  • Antigen symbol:
  • TAF11
  • TAF11
  • Antigen synonyms:
  • TAFII-28|TAF11_HUMAN|PRO2134|Transcription initiation factor TFIID subunit 11|TAF11 RNA polymerase II TATA box binding protein (TBP) associated factor 28 kD|TAF2I|p30 beta|TAF11|TAFII 28|TAF(II)28|TFIID subunit p30 beta|Transcription initiation factor TFIID 28 kDa subunit|MGC:15243|TFIID subunit p30-beta|TAF|TAFII28
  • TAFII-28|TAF11_HUMAN|PRO2134|Transcription initiation factor TFIID subunit 11|TAF11 RNA polymerase II TATA box binding protein (TBP) associated factor 28 kD|TAF2I|p30 beta|TAF11|TAFII 28|TAF(II)28|TFIID subunit p30 beta|Transcription initiation factor TFIID 28 kDa subunit|MGC:15243|TFIID subunit p30-beta|TAF|TAFII28
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q15544
  • Q15544
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Fusion protein of Human TAF11
  • Fusion protein of Human TAF11
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together