Anti-FOXO4 (acetyl-Lys189) Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
WB:1:500-2000, ELISA:1:10000-20000
Type:
Antigen: FOXO4 (acetyl-Lys189)
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77705-347
- 77712-997
- Antigen symbol:
- FOXO4 (acetyl-Lys189)
- FOXO4 (acetyl-Lys189)
- Antigen synonyms:
- Fork head domain transcription factor AFX1|MLLT7|FOXO 4|ALL1-fused gene from X chromosome|RGD1561201|Myeloid/lymphoid or mixed lineage leukemia (trithorax homolog|Myeloid/lymphoid or mixed lineage leukemia|AFX1|translocated to|7|MGC120490|MGC117660|Forkhead box O4|Mixed lineage leukemia|Drosophila)|Forkhead box protein O4|Foxo4|AFX|Afxh|FOXO4_HUMAN
- Fork head domain transcription factor AFX1|MLLT7|FOXO 4|ALL1-fused gene from X chromosome|RGD1561201|Myeloid/lymphoid or mixed lineage leukemia (trithorax homolog|Myeloid/lymphoid or mixed lineage leukemia|AFX1|translocated to|7|MGC120490|MGC117660|Forkhead box O4|Mixed lineage leukemia|Drosophila)|Forkhead box protein O4|Foxo4|AFX|Afxh|FOXO4_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P98177
- P98177
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic Acetyl peptide from Human protein at AA range: 189
- Synthetic Acetyl peptide from Human protein at AA range: 189
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together