Anti-RCBTB2 Rabbit Polyclonal Antibody
Catalog # 77688-559
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:RCBTB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O95199
- Antigen synonyms:RCC1 and BTB domain containing protein 2|RCC1 like G exchanging factor RLG|Chromosome condensation 1 like|RCBT2_HUMAN|Regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2|Regulator of chromosome condensation (RCC1) and BTB domain containing protein 2|CHC1L|CHC1 L|RCC1 like G exchanging factor|Regulator of chromosome condensation and BTB domain containing protein 2|RCBT2|RCC1 and BTB domain-containing protein 2|RLG|Rcbtb2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human RCC1 and BTB domain-containing protein 2 protein (1-200AA)
- Purification:>95%, Protein G purified
- Cat. no.:77688-559
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: RCBTB2
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat