Anti-MAGEA10 Rabbit Polyclonal Antibody
77688-208
New Product
- Antigen symbol:MAGEA10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P43363
- Antigen synonyms:Cancer/testis antigen 1.10|CT1.10|MAGE10Melanoma-associated antigen 10|MAGE-10 antigen|MAGEA10
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Melanoma-associated antigen 10 protein (2-225AA)
- Purification:>95%, Protein G purified
- Cat. no.:77688-208
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200
Type:
Antigen: MAGEA10
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human