Anti-HIST1H2AG (acetyl-K36) Rabbit Polyclonal Antibody
77687-869
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen symbol:HIST1H2AG (acetyl-K36)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Environmentally Preferable:
- ChIP:Yes
- Form:Liquid
- Gene ID:P0C0S8
- Antigen synonyms:HIST1H2AMHistone H2A type 1|HIST1H2AL|H2A.1|H2AC17|H2AFI|H2AC11|H2AC16|Histone H2A/ptl|H2AFN|HIST1H2AG|HIST1H2AK|H2AFD|H2AC13|H2AC15|HIST1H2AI|H2AFC|H2AFP
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around site of Acetyl-Lys (36) derived from Human Histone H2A type 1
- Purification:Antigen affinity purified
- Cat. no.:77687-869
- Packaging:Vial
Recommended dilution: WB:1:200-1:2000, ICC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: HIST1H2AG (acetyl-K36)
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human