Anti-MAEA Rabbit Polyclonal Antibody
Catalog # 77699-435
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MAEA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q7L5Y9
- Antigen synonyms:Cell proliferation-inducing gene 5 protein|1110030D19Rik|GID complex subunit 9 FYV10 homolog|PIG5|MGC93683|HLC-10|Macrophage erythroblast attacher|Proliferation inducing gene 5|Cell proliferation inducing gene 5 protein|EMLP|MAEA|Lung cancer related protein 10|Human lung cancer oncogene 10 protein|HLC 10|GID9|HLC10|Erythroblast macrophage protein|MAEA_HUMAN|EMP
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Emp.
- Purification:Antigen affinity purified
- Cat. no.:77699-435
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MAEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat