Anti-EEF1G Rabbit Polyclonal Antibody
77699-408
New Product
- Antigen symbol:EEF1G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P26641
- Antigen synonyms:eEF 1B gamma|EF 1 gamma|EF 1G|MGC144723|MGC94929|MGC103354|Pancreatic tumor related protein|Eukaryotic translation elongation factor 1 gamma|EEF 1G|EEF1G|Elongation factor 1 gamma|eEF-1B gamma|GIG35|EF1G_HUMAN|EF1G|Elongation factor 1-gamma|2610301D06Rik|AA407312|EF-1-gamma|GIG 35|MGC144724|Translation elongation factor eEF 1 gamma chain|PRO1608|EF1 gamma|MGC114210
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human EF-1 γ.
- Purification:Antigen affinity purified
- Cat. no.:77699-408
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: EEF1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat