Anti-DCLK3 Rabbit Polyclonal Antibody
Catalog # 77699-331
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:DCLK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9C098
- Antigen synonyms:CLICK I and II related|hCG_16695|Click I II related|RGD1309232|BC056929|DCK3|Doublecortin domain-containing protein 3C|Doublecortin-like kinase 3|Dclk3|Dcamkl3|KIAA1765|Doublecortin and CaM kinase like 3|C730036H08|Doublecortin-like and CAM kinase-like 3|DCDC3C|CLr|CLR|Serine/threonine-protein kinase DCLK3|DCLK3_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human DCAMKL3.
- Purification:Antigen affinity purified
- Cat. no.:77699-331
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: DCLK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human