Anti-CNGA2 Rabbit Polyclonal Antibody
Catalog # 77699-197
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CNGA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q16280
- Antigen synonyms:CNG channel alpha-2|FLJ46312|CNG-2|OCNC1|Cyclic nucleotide gated olfactory channel|CNCA|CNCG2|CNG 2|CNG2|Cyclic nucleotide-gated cation channel 2|Cyclic nucleotide gated cation channel 2|Cyclic nucleotide gated channel alpha 2|OCNCALPHA|CNG channel alpha 2|CNGA 2|OCNCa|CNCA1|Cyclic nucleotide-gated olfactory channel|Cyclic nucleotide-gated channel alpha-2|CNGA2_HUMAN|Cnga2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CNG-2.
- Purification:Antigen affinity purified
- Cat. no.:77699-197
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: CNGA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat