Anti-CEP290 Rabbit Polyclonal Antibody
77699-138
New Product
- Antigen symbol:CEP290
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O15078
- Antigen synonyms:Cep290|type 4|3H11AG|Prostate cancer antigen T21|Cancer/testis antigen 87|Bardet-Biedl syndrome 14 protein|SLSN6 1|MKS4|POC3 centriolar protein homolog|Nephrocystin 6|Tumor antigen se2-2|Centrosomal protein 290|SLSN6|CTCL tumor antigen se2 2|CE290_HUMAN|JBTS6|Centrosomal protein of 290 kDa|FLJ21979|rd16|Nephrocystin-6|LCA10|Meckel syndrome|FLJ13615|BBS14|2|KIAA0373|POC3|NPHP6|5|JBTS5|Centrosomal protein 290kDa|CT87|Monoclonal 3H11 antigen
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CEP290.
- Purification:Antigen affinity purified
- Cat. no.:77699-138
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: CEP290
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse