Anti-CD2BP2 Rabbit Polyclonal Antibody
Catalog # 77699-095
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:CD2BP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O95400
- Antigen synonyms:CD2 (cytoplasmic tail) binding protein 2 |U5-52K|FWP010|U5 snRNP 52K protein|Cd2bp2|CD2 cytoplasmic domain-binding protein 2|CD2 antigen cytoplasmic tail-binding protein 2|CD2 antigen (cytoplasmic tail) binding protein 2|Snu40|CD2 tail-binding protein 2|LIN1|CD2B2_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human CD2BP2.
- Purification:Antigen affinity purified
- Cat. no.:77699-095
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: CD2BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey