Anti-VWA5A Rabbit Polyclonal Antibody
Catalog # 77698-958
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:VWA5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O00534
- Antigen synonyms:BCSC1|BCSC-1|VWA5A|Loss of heterozygosity 11 chromosomal region 2 gene A protein|LOH11CR2A|von Willebrand factor A domain-containing protein 5A|Breast cancer suppressor candidate 1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human BCSC-1.
- Purification:Antigen affinity purified
- Cat. no.:77698-958
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: VWA5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human