Anti-ATP5C1 Rabbit Polyclonal Antibody
77698-919
New Product
- Antigen symbol:ATP5C1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P36542
- Antigen synonyms:ATP5C1|ATPG_HUMAN|F-ATPase gamma subunit|ATP synthase gamma chain|ATP5C 1|ATP5C|ATP5CL1|gamma polypeptide 1|mitochondrial|ATP synthase|gamma subunit|Mitochondrial ATP synthase|gamma subunit 1|F ATPase gamma subunit|H+ transporting|ATP synthase subunit gamma|mitochondrial F1 complex
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ATP5C1.
- Purification:Antigen affinity purified
- Cat. no.:77698-919
- Packaging:Vial
Type:
Antigen: ATP5C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat