Anti-ACACA Rabbit Polyclonal Antibody
Catalog # 77698-787
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ACACA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q13085
- Antigen synonyms:Acetyl CoA carboxylase alpha |OTTHUMP00000240532|ACC1|ACACB|ACACA_HUMAN|Acetyl CoA carboxylase 2|ACC alpha|ACCA|ACC-alpha|Biotin carboxylase|Acetyl Coenzyme A carboxylase beta|ACC2|ACAC|OTTHUMP00000164069|Acetyl Coenzyme A carboxylase alpha|Acetyl CoA carboxylase 1|OTTHUMP00000164076|COA2|Acetyl CoA carboxylase beta|OTTHUMP00000164070|ACC|ACACA|ACC beta |HACC275|COA1|ACCB
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human ACCα around the non-phosphorylation site of S80.
- Purification:Antigen affinity purified
- Cat. no.:77698-787
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: ACACA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat