Anti-TRIM25 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77711-614
- 77711-615
- Antigen symbol:
- TRIM25
- TRIM25
- Antigen synonyms:
- RING finger protein 147|Tripartite motif-containing protein 25|ZNF 147|Tripartite motif containing 25|Zinc finger protein 147|Z147|ZNF147|Ubiquitin/ISG15-conjugating enzyme TRIM25|Estrogen-responsive finger protein|TRI25|Tripartite motif containing protein 25|Zinc finger protein 147 (estrogen responsive finger protein)|EFP|RNF 147|RNF147|Estrogen responsive finger protein|E3 ubiquitin/ISG15 ligase TRIM25|Tripartite motifcontaining25|Tripartite motif protein TRIM25|TRIM 25|Trim25|Zinc finger protein-147|TRI25_HUMAN
- RING finger protein 147|Tripartite motif-containing protein 25|ZNF 147|Tripartite motif containing 25|Zinc finger protein 147|Z147|ZNF147|Ubiquitin/ISG15-conjugating enzyme TRIM25|Estrogen-responsive finger protein|TRI25|Tripartite motif containing protein 25|Zinc finger protein 147 (estrogen responsive finger protein)|EFP|RNF 147|RNF147|Estrogen responsive finger protein|E3 ubiquitin/ISG15 ligase TRIM25|Tripartite motifcontaining25|Tripartite motif protein TRIM25|TRIM 25|Trim25|Zinc finger protein-147|TRI25_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14258
- Q14258
- Isotype:
- IgG
- IgG
- Immunogen:
- Synthetic peptide of Human TRIM25
- Synthetic peptide of Human TRIM25
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together