Anti-TMPRSS11E Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, IHC:1:30-1:150
Type:
Antigen: TMPRSS11E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77712-032
- 77712-031
- Antigen symbol:
- TMPRSS11E
- TMPRSS11E
- Antigen synonyms:
- UNQ742/PRO1461|EC 3.4.21.-|Transmembrane protease serine 11E2) [Cleaved into: Transmembrane protease serine 11E non-catalytic chain|TMPRSS11E2|Transmembrane protease serine 11E|Transmembrane protease serine 11E catalytic chain]|DESC1|TMPRSS11E|Serine protease DESC1
- UNQ742/PRO1461|EC 3.4.21.-|Transmembrane protease serine 11E2) [Cleaved into: Transmembrane protease serine 11E non-catalytic chain|TMPRSS11E2|Transmembrane protease serine 11E|Transmembrane protease serine 11E catalytic chain]|DESC1|TMPRSS11E|Serine protease DESC1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UL52
- Q9UL52
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TMPRSS11E
- Fusion protein of Human TMPRSS11E
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together