Anti-STK36 Rabbit Polyclonal Antibody
Catalog # 77702-598
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:STK36
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9NRP7
- Antigen synonyms:STK36_HUMAN|FU|Serine/threonine protein kinase 36 fused homolog|STK36|KIAA1278|Serine/threonine-protein kinase 36|Fused homolog|Serine/threonine kinase 36, fused homolog
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human STK36.
- Purification:Antigen affinity purified
- Cat. no.:77702-598
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: STK36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse