Anti-polA Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: polA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Thermus aquaticus
Specifications
- Catalog No:
- 77711-252
- 77711-251
- Antigen symbol:
- polA
- polA
- Antigen synonyms:
- thermostable|Taq polymerase 1|EC 2.7.7.7|DNA polymerase I|polA|pol1
- thermostable|Taq polymerase 1|EC 2.7.7.7|DNA polymerase I|polA|pol1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Thermus aquaticus
- Thermus aquaticus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19821
- P19821
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Thermus aquaticus DNA polymerase I, thermostable protein (310-832AA)
- Recombinant Thermus aquaticus DNA polymerase I, thermostable protein (310-832AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together