Anti-PIP4K2A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: PIP4K2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77712-100
- 77712-101
- Antigen symbol:
- PIP4K2A
- PIP4K2A
- Antigen synonyms:
- 4 5 kinase B isoform|Diphosphoinositide kinase 2 alpha|1 phosphatidylinositol 4 phosphate kinase|Phosphatidylinositol 4 phosphate 5 kinase type II alpha|PtdIns|PIP5K2|PIP5KII alpha|PtdIns(4)P-5-kinase B isoform|PtdIns(5)P-4-kinase isoform 2-alpha|PIP4K2A|PIP5K2 alpha|PIP5KIII|PI5P4KA|Phosphatidylinositol-5-phosphate 4-kinase type-2 alpha|4 5 kinase isoform 2 alpha|PI42A_HUMAN|PIP5KII-alpha|PIPK|PtdIns(4)P-5-kinase C isoform|Phosphatidylinositol 4 phosphate 5 kinase type 2 alpha|1 phosphatidylinositol 4 phosphate 5 kinase 2 alpha|PI(5)P 4-kinase type II alpha|Phosphatidylinositol-5-phosphate 4-kinase type II alpha|4 5 kinase C isoform|1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha|Type II phosphatidylinositol 4 phosphate 5 kinase 53 K isoform|FLJ13267|PIP4KII-alpha|Diphosphoinositide kinase 2-alpha|PIP5KIIA|1 phosphatidylinositol 4 phosphate 5 kinase|PIP5K2A
- 4 5 kinase B isoform|Diphosphoinositide kinase 2 alpha|1 phosphatidylinositol 4 phosphate kinase|Phosphatidylinositol 4 phosphate 5 kinase type II alpha|PtdIns|PIP5K2|PIP5KII alpha|PtdIns(4)P-5-kinase B isoform|PtdIns(5)P-4-kinase isoform 2-alpha|PIP4K2A|PIP5K2 alpha|PIP5KIII|PI5P4KA|Phosphatidylinositol-5-phosphate 4-kinase type-2 alpha|4 5 kinase isoform 2 alpha|PI42A_HUMAN|PIP5KII-alpha|PIPK|PtdIns(4)P-5-kinase C isoform|Phosphatidylinositol 4 phosphate 5 kinase type 2 alpha|1 phosphatidylinositol 4 phosphate 5 kinase 2 alpha|PI(5)P 4-kinase type II alpha|Phosphatidylinositol-5-phosphate 4-kinase type II alpha|4 5 kinase C isoform|1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha|Type II phosphatidylinositol 4 phosphate 5 kinase 53 K isoform|FLJ13267|PIP4KII-alpha|Diphosphoinositide kinase 2-alpha|PIP5KIIA|1 phosphatidylinositol 4 phosphate 5 kinase|PIP5K2A
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48426
- P48426
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PIP4K2A
- Fusion protein of Human PIP4K2A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together